Anti-GRM6 Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A23968-100UG A23968-50UG
77739-067EA 521.4 CAD
77739-067 77856-482
Anti-GRM6 Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
0

Type:
Antigen: GRM6
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR