Anti-FRS2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A22456-100UG A22456-50UG
77737-643EA 521.4 CAD
77737-643 77855-618
Anti-FRS2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: FRS2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol FRS2
Environmentally Preferable
Antigen synonyms FGFR signaling adaptor SNT|FGFR signalling adaptor|FGFR substrate 2|FGFR-signaling adaptor SNT|Fibroblast growth factor receptor substrate 2|FRS 2|FRS2_HUMAN|FRS2A|FRS2alpha|SNT 1|SNT|SNT-1|SNT1|Suc 1 Associated Neurotrophic Factor Target|Suc1 associated neurotrophic factor target 1|Suc1-associated neurotrophic factor target 1
Clonality Recombinant Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human
Host Rabbit
Gene ID Q8WU20
Isotype IgG
ELISA Yes
Immunogen Recombinant Human Fibroblast growth factor receptor substrate 2 protein (116-387AA)
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR