Anti-HIST1H2BB Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A24831-100UL A24831-50UL
77739-816EA 553 CAD
77739-816 77857-046
Anti-HIST1H2BB Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ICC: 1:20 to 1:200, IF: 1:50 to 1:200

Type:
Antigen: HIST1H2BB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR