Anti-Gpi Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A23644-100UG A23644-50UG
77738-751EA 521.4 CAD
77738-751 77856-312
Anti-Gpi Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: Gpi
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR