Anti-MEF2A Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A28406-100UL A28406-50UL
77742-979EA 521.4 CAD
77742-979 77859-639
Anti-MEF2A Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:100 to 1:1000

Type:
Antigen: MEF2A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR