Anti-GLYCTK Rabbit Recombinant polyclonal Antibody

Supplier: AFG Bioscience
A23356-100UL A23356-50UL
77738-479EA 521.4 CAD
77738-479 77856-137
Anti-GLYCTK Rabbit Recombinant polyclonal Antibody
Antibodies
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200

Type:
Antigen: GLYCTK
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR