Anti-HIST1H3A Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A24942-100UL A24942-50UL
77739-887EA 553 CAD
77739-887 77857-157
Anti-HIST1H3A Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:200-1:2000, IF:1:1-1:10, IP:1:200-1:2000

Type:
Antigen: HIST1H3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR