Anti-dsdA Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A52378-100UG A52378-50UG
77818-850EA 521.4 CAD
77818-850 77818-851
Anti-dsdA Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: dsdA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Salmonella heidelberg
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR