Anti-FAM96A Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A68266-100UG A68266-50UG
77838-401EA 521.4 CAD
77838-401 77838-402
Anti-FAM96A Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: FAM96A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR