Anti-STXBP1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A35929-100UG A35929-50UG
77801-488EA 521.4 CAD
77801-488 77801-489
Anti-STXBP1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200

Type:
Antigen: STXBP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR