Anti-KIR2DS3 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A54213-100UG A54213-50UG
77822-069EA 521.4 CAD
77822-069 77822-070
Anti-KIR2DS3 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: KIR2DS3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR