Anti-PDE6H Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A54722-100UG A54722-50UG
77823-030EA 521.4 CAD
77823-030 77823-031
Anti-PDE6H Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: PDE6H
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR