Anti-ASNS Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A53826-100UL A53826-50UL
77821-347EA 521.4 CAD
77821-347 77821-348
Anti-ASNS Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA:1:1000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:50 to 1:200

Type:
Antigen: ASNS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR