Anti-RNF26 Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A33368-100UG A33368-50UG
77797-542EA 521.4 CAD
77797-542 77797-543
Anti-RNF26 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies

Type:
Antigen: OXCT1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human


Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR