Anti-GPAM Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A68891-50UG A68891-100UG
77839-578EA 355.5 CAD
77839-578 77839-577
Anti-GPAM Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: GPAM
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR