Anti-AGXT2 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A67483-100UG A67483-50UG
77836-928EA 521.4 CAD
77836-928 77836-929
Anti-AGXT2 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: AGXT2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR