Anti-GATD3A Rabbit Recombinant Antibody (HRP (Horseradish Peroxidase))

Supplier: AFG Bioscience
A31389-50UG A31389-100UG
77794-414EA 355.5 CAD
77794-414 77794-413
Anti-GATD3A Rabbit Recombinant Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type:
Antigen: GATD3A
Clonality: Recombinant
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR