Anti-BDNF Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A54691-100UL A54691-50UL
77822-976EA 521.4 CAD
77822-976 77822-977
Anti-BDNF Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA:1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:10 to 1:50

Type:
Antigen: BDNF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR