Anti-GK3P Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A54443-100UG A54443-50UG
77822-504EA 521.4 CAD
77822-504 77822-505
Anti-GK3P Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: GK3P
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR