Anti-SLC6A8 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A40569-100UG A40569-50UG
77807-857EA 521.4 CAD
77807-857 77807-858
Anti-SLC6A8 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: SLC6A8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR