Anti-GCY1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A50424-100UG A50424-50UG
77815-264EA 521.4 CAD
77815-264 77815-265
Anti-GCY1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: GCY1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Saccharomyces cerevisiae
New Product
Order Now

SPECIFICATIONS

Antigen symbol GCY1
Antigen name Recombinant Saccharomyces cerevisiae Glycerol 2-dehydrogenase protein (1-312AA)
Environmentally Preferable
Antigen synonyms GCY1|GCY|YOR120W|O31567|YOR3269WGlycerol 2-dehydrogenase|NADP(+)|EC 1.1.1.156|Galactose-inducible crystallin-like protein 1
Clonality Recombinant Polyclonal
Conjugation Unconjugated
Reactivity S. cerevisiae
Host Rabbit
Gene ID P14065
Isotype IgG
ELISA Yes
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR