Anti-NFKBIL1 Rabbit Recombinant Antibody (Biotinylated)

Supplier: AFG Bioscience
A70515-100UG A70515-50UG
77842-644EA 521.4 CAD
77842-644 77842-645
Anti-NFKBIL1 Rabbit Recombinant Antibody (Biotinylated)
Antibodies
Type:
Antigen: NFKBIL1
Clonality: Recombinant
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR