Anti-Toxin zeta Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A51700-100UG A51700-50UG
77817-671EA 521.4 CAD
77817-671 77817-672
Anti-Toxin zeta Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: Toxin zeta
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Streptococcus agalactiae
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR