Anti-DPH2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: AFG Bioscience
A66287-100UG A66287-50UG
77834-680EA 521.4 CAD
77834-680 77834-681
Anti-DPH2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type:
Antigen: DPH2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol DPH2
Antigen name 2-(3-amino-3-carboxypropyl)histidine synthase subunit 2 protein (1-489AA)
Environmentally Preferable
Antigen synonyms Diphthamide biosynthesis like protein 2| Diphthamide biosynthesis protein 2| Diphthamide biosynthesis protein 2 homolog like 2| Diphthamide biosynthesis protein 2 homolog-like 2| Diptheria toxin resistance protein required for diphthamide biosynthesis like 2| Diptheria toxin resistance protein required for diphthamide biosynthesis like 2 S. cerevisiae| DPH 2| DPH 2 homolog| DPH2| DPH2 homolog| DPH2 homolog S. cerevisiae| DPH2 like 2| DPH2 like 2 S. cerevisiae| DPH2-like 2| DPH2_HUMAN| DPH2L2| HsDph2| OTTHUMP00000010007| Protein DPH2 homolog
Clonality Recombinant Polyclonal
Conjugation HRP (Horseradish Peroxidase)
Reactivity Human
Host Rabbit
Gene ID Q9BQC3
Isotype IgG
ELISA Yes
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR