Anti-PGS1 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A56283-100UG A56283-50UG
77825-978EA 521.4 CAD
77825-978 77825-979
Anti-PGS1 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: PGS1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol PGS1
Antigen name CDP-diacylglycerol--glycerol-3-phosphate 3-phosphatidyltransferase, mitochondrial protein (110-400AA)
Environmentally Preferable
Antigen synonyms CDP diacylglycerol glycerol 3 phosphate 3 phosphatidyltransferase mitochondrial|CDP-diacylglycerol--glycerol-3-phosphate 3-phosphatidyltransferase|HCG1642326, isoform CRA_b|mitochondrial|PGP synthase 1|PGPS1_HUMAN|PGS1|PGS1 protein Fragment|Phosphatidylglycerophosphate synthase 1
Clonality Recombinant Polyclonal
Conjugation HRP (Horseradish Peroxidase)
Reactivity Human
Host Rabbit
Gene ID Q32NB8
Isotype IgG
ELISA Yes
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR