Anti-AGPAT1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A66445-100UL A66445-50UL
77834-978EA 521.4 CAD
77834-978 77834-979
Anti-AGPAT1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: AGPAT1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR