Anti-BICC1 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A68269-100UG A68269-50UG
77838-407EA 521.4 CAD
77838-407 77838-408
Anti-BICC1 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: BICC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR