Anti-CAPRIN2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A58088-100UL A58088-50UL
77829-347EA 521.4 CAD
77829-347 77829-348
Anti-CAPRIN2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200

Type:
Antigen: CAPRIN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR