Anti-ZNF333 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A38895-100UG A38895-50UG
77805-330EA 521.4 CAD
77805-330 77805-331
Anti-ZNF333 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: ZNF333
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR