Anti-AIP Rabbit Recombinant Antibody

Supplier: AFG Bioscience
A52506-100UG A52506-50UG
77819-081EA 521.4 CAD
77819-081 77819-082
Anti-AIP Rabbit Recombinant Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: AIP
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol AIP
Environmentally Preferable
Antigen name AH receptor-interacting protein (105-294AA)
Antigen synonyms AH receptor interacting protein|AH receptor-interacting protein|AIP|AIP_HUMAN|ARA 9|ARA9|Aryl hydrocarbon receptor interacting protein|Aryl-hydrocarbon receptor-interacting protein|fa03h10|FKBP 16|FKBP 37|FKBP16|FKBP37|HBV X associated protein 2|HBV X associated protein|HBV X-associated protein 2|HBVX associated protein|Immunophilin homolog ARA 9|Immunophilin homolog ARA9|SMTPHN|XAP 2|XAP-2|XAP2
Clonality Recombinant Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID O00170
Isotype IgG
ELISA Yes
ImmunoChemistry Yes
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR