Anti-ZNF432 Rabbit Recombinant Antibody

Supplier: AFG Bioscience
A38958-100UG A38958-50UG
77805-445EA 521.4 CAD
77805-445 77805-446
Anti-ZNF432 Rabbit Recombinant Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: ZNF432
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR