Anti-CEBPE Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A54823-100UG A54823-50UG
77823-219EA 521.4 CAD
77823-219 77823-220
Anti-CEBPE Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:200 to 1:2000

Type:
Antigen: CEBPE
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR