Anti-POLD2 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A31565-50UG A31565-100UG
77794-694EA 355.5 CAD
77794-694 77794-693
Anti-POLD2 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: POLD2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR