Anti-CXCL1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A41735-100UG A41735-50UG
77809-456EA 521.4 CAD
77809-456 77809-457
Anti-CXCL1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: CXCL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR