Anti-PROC Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A32070-100UG A32070-50UG
77795-481EA 521.4 CAD
77795-481 77795-482
Anti-PROC Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: PROC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol PROC
Antigen name Vitamin K-dependent protein C protein (288-409AA)
Environmentally Preferable
Antigen synonyms Activation peptide|Anticoagulant protein C|APC|Autoprothrombin IIA|Blood coagulation factor XIV|EC 3.4.21.69|PC|proC|PROC_HUMAN|PROC1|Protein C (inactivator of coagulation factors Va and VIIIa)|THPH3|THPH4|Vitamin K dependent protein C
Clonality Recombinant Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human
Host Rabbit
Gene ID P04070
Isotype IgG
ELISA Yes
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR