Anti-PPP1CC Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A31812-100UG A31812-50UG
77795-074EA 521.4 CAD
77795-074 77795-075
Anti-PPP1CC Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: PPP1CC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR