Anti-SNX29 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A49638-100UL A49638-50UL
77813-851EA 521.4 CAD
77813-851 77813-852
Anti-SNX29 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:20 to 1:100

Type:
Antigen: SNX29
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR