Anti-Allergen Ara h 1, clone P17 Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A50760-100UG A50760-50UG
77815-900EA 521.4 CAD
77815-900 77815-901
Anti-Allergen Ara h 1, clone P17 Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: Allergen Ara h 1, clone P17
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Peanut
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR