Anti-TGM4 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A36515-100UG A36515-50UG
77802-439EA 521.4 CAD
77802-439 77802-440
Anti-TGM4 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:200 to 1:500, IF:1:50 to 1:200

Type:
Antigen: TGM4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR