Anti-C1QTNF9B Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A52334-100UG A52334-50UG
77818-774EA 521.4 CAD
77818-774 77818-775
Anti-C1QTNF9B Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: C1QTNF9B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR