Anti-PGP Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A31092-100UG A31092-50UG
77793-940EA 521.4 CAD
77793-940 77793-941
Anti-PGP Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: PGP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol PGP
Environmentally Preferable
Antigen name Glycerol-3-phosphate phosphatase protein (140-300AA)
Antigen synonyms PGP|PGP_HUMAN|PGPase|Phosphoglycolate phosphatase
Clonality Recombinant Polyclonal
Conjugation Biotin
Reactivity Human
Host Rabbit
Gene ID A6NDG6
Isotype IgG
ELISA Yes
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
Storage temperature Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR