Anti-VNN1 Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A38327-100UG A38327-50UG
77804-398EA 521.4 CAD
77804-398 77804-399
Anti-VNN1 Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: VNN1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR