Anti-GBE1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A58792-100UL A58792-50UL
77830-684EA 521.4 CAD
77830-684 77830-685
Anti-GBE1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200

Type:
Antigen: GBE1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR