Anti-CARD14 Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A70045-100UG A70045-50UG
77841-755EA 521.4 CAD
77841-755 77841-756
Anti-CARD14 Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: CARD14
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR