Anti-EDAR Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A43303-100UL A43303-50UL
77812-815EA 521.4 CAD
77812-815 77812-816
Anti-EDAR Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:35 to 1:150

Type:
Antigen: EDAR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR