Anti-TPK1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A37291-100UG A37291-50UG
77811-371EA 521.4 CAD
77811-371 77803-371
Anti-TPK1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IF: 1:50 to 1:200

Type:
Antigen: TPK1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR