Anti-KCNJ11 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A55698-100UG A55698-50UG
77824-877EA 521.4 CAD
77824-877 77824-878
Anti-KCNJ11 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500 to 1:5000, IHC:1:20 to 1:200

Type:
Antigen: KCNJ11
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR