Anti-ABHD10 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A67270-100UG A67270-50UG
77836-531EA 521.4 CAD
77836-531 77836-532
Anti-ABHD10 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: ABHD10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR