Anti-IFT80 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A70485-50UG A70485-100UG
77842-588EA 355.5 CAD
77842-588 77842-587
Anti-IFT80 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: IFT80
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR