Anti-EHD2 Rabbit Recombinant Antibody (Biotinylated)

Supplier: AFG Bioscience
A68562-100UG A68562-50UG
77838-955EA 521.4 CAD
77838-955 77838-956
Anti-EHD2 Rabbit Recombinant Antibody (Biotinylated)
Antibodies
Type:
Antigen: EHD2
Clonality: Recombinant
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR